Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID5B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen ARID5B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16916920
SDP
View Documents
Novus Biologicals
NBP16916920UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP169169
SDP
View Documents
Novus Biologicals
NBP169169
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

ARID5B Polyclonal specifically detects ARID5B in Human samples. It is validated for Western Blot.
Specifications

Specifications

ARID5B
Polyclonal
Rabbit
Q14865
84159
Synthetic peptides corresponding to ARID5B (AT rich interactive domain 5B (MRF1-like)) The peptide sequence was selected from the C terminal of ARID5B (NP_115575). Peptide sequence: VSPLDPSKEVSGKEKASEQESEGSKAAHGGHSGGGSEGHKLPLSSPIFPG.
Primary
Western Blot
Unconjugated
RUO
ARID domain-containing protein 5B, AT rich interactive domain 5B (MRF1-like), DESRTAT-rich interactive domain-containing protein 5B, FLJ21150, Modulator recognition factor 2, modulator recognition factor 2 (MRF2), MRF-2, MRF2MRF1-like protein
ARID5B
IgG
132 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.