Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARID5B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$416.50 - $697.00
Specifications
Antigen | ARID5B |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ARID5B Polyclonal specifically detects ARID5B in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ARID5B | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
84159 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TSKYPSRDMYRESENSSFPSHRHQEKLHVNYLTSLHLQDKKSAAAEAPTDDQPTDLSLPKNPHKPTGKVLGLAHSTTGPQESKGISQFQVLGSQSRDCHPKACRVSPMTMSGPKKYPESLSRSGKPHHVRLENFRKME | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
ARID domain-containing protein 5B, AT rich interactive domain 5B (MRF1-like), DESRTAT-rich interactive domain-containing protein 5B, FLJ21150, Modulator recognition factor 2, modulator recognition factor 2 (MRF2), MRF-2, MRF2MRF1-like protein | |
ARID5B | |
IgG | |
Affinity Purified | |
Specificity of human ARID5B antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title