Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARIH1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155039 Supplier Novus Biologicals Supplier No. NBP155039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARIH1 Polyclonal specifically detects ARIH1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ARIH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9Y4X5
Gene Alias ARI-1, ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1(Drosophila), ariadne, Drosophila, homolog of, ARIMOP-6, FLJ20329, FLJ93118, H7-AP2, HARI, Monocyte protein 6, MOP6, protein ariadne-1 homolog, UbcH7-binding protein, UBCH7BPariadne (Drosophila) homolog, ubiquitin-conjugating enzyme E2-binding protein1,HHARIDKFZp686O13120, UbcM4-interacting protein, Ubiquitin-conjugating enzyme E2-binding protein 1
Gene Symbols ARIH1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ARIH1(ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1 (Drosophila)) The peptide sequence was selected from the C terminal of ARIH1 (NP_005735). Peptide sequence ATEVLSGYLERDISQDSLQDIKQKVQDKYRYCESRRRVLLQHVHEGYEKD The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 61 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25820
Reconstitution Reconstitute in 100μL distilled water to a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.