Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Arium™ Mini Extend Ultrapure Water System
Shop All Sartorius Products
Click to view available options
Model:
Arium Mini Essential with UV
Arium Mini Essential without UV
Arium Mini Plus with UV
Arium Mini Plus without UV
Description
- Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
- Compact: Space-saving with a width of only 28 cm (+ 9 cm for versions with attached dispensing unit)
- Intuitive: Color touch display, with direct access to all important information and dispensing options
- Innovative: Depending on the type of system, our unique Bagtank technology saves time as no intensive tank cleaning with hazardous substances is required
- Reliable: Delivers consistently high water quality for reliable and reproducible results

Specifications
Specifications
Bacterial Content | <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) |
Daily Output | 1 to 10 L |
Endotoxin Concentration | <0.001 EU/mL with a terminal ultrafiltration cartridge |
Feedwater Connector | 3/8 in. |
Feed Water Monitoring | No |
Feedwater Source | Distilled, DI, RO, EDI |
Flow Rate | 1 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 MΩ x cm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction