Learn More
Eisco™ Arkose Sandstone Specimen (Sedimentary Rock)
SCI_ED
Catalog No. S24716 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24716 1 Ea.
S26676 10/Pk.
S24717 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24716 Supplier Eisco™ Supplier No. ESNG0028
Add to Cart
Add to Cart
Arkose Sandstone is clastic sedimentary rock made up of angular, poorly-sorted sand-size grains.
  • Small, angular grains indicate deposition in ephemeral river system near to clast source
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Sedimentary Rock
For Use With (Application) Lessons in identifying provenance and interpreting depositional environment
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Arkose Sandstone
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov