Learn More
Eisco™ Arkose Sandstone Specimen (Sedimentary Rock)
Arkose Sandstone is clastic sedimentary rock made up of angular, poorly-sorted sand-size grains.
$3.18 - $54.10
Specifications
Product Type | Sedimentary Rock, Arkose Sandstone |
---|---|
Specimen Size | 2 to 3 cm |
Description
- Small, angular grains indicate deposition in ephemeral river system near to clast source
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Sedimentary Rock, Arkose Sandstone | |
2 to 3 cm |