Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ARL 17477 dihydrochloride
GREENER_CHOICE

Catalog No. 331910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-191-0 10 mg
33-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-191-0 Supplier Tocris Bioscience™ Supplier No. 3319/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective nNOS inhibitor

Selective neuronal nitrogen oxide synthase (nNOS) inhibitor (IC50 values are 1 and 17 μM for nNOS and endothelial NOS respectively). Reduces ischemic cell damage after middle cerebral artery (MCA) occlusion in rats. Displays a synergistic neuroprotective effect when combined with either an NMDA or AMPA receptor antagonist.

Chemical Identifiers

CAS 866914-87-6
Molecular Formula C20H22Cl3N3S

Specifications

Quantity 10 mg
Formula Weight 442.83
Purity >99%
Molecular Weight (g/mol) 442.827
Chemical Name or Material ARL 17477 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.