Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL11 Antibody (OTI1A5), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N272348P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272348P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272348P594 Supplier Novus Biologicals Supplier No. NBP272348PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARL11 Monoclonal specifically detects ARL11 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ARL11
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1A5
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADP-ribosylation factor-like 11, ADP-ribosylation factor-like protein 11, ADP-ribosylation factor-like tumor suppressor protein 1, ARLTS1FLJ33930, MGC17429
Gene Symbols ARL11
Host Species Mouse
Immunogen Full length recombinant protein of human ARL11 (NP_612459) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 115761.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.