Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL14EP Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB332076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332076 Supplier Novus Biologicals Supplier No. NBP297378MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARL14EP Polyclonal antibody specifically detects ARL14EP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ARL14EP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosylation factor-like 14 effector protein, ARF7EP, C11orf46, chromosome 11 open reading frame 46, dJ299F11.1, FLJ38968, hypothetical protein LOC120534
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ARL14EP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120534
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.