Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARL14EP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | C11orf46 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15647820
![]() |
Novus Biologicals
NBP15647820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156478
![]() |
Novus Biologicals
NBP156478 |
100 μL |
Each for $487.50
|
|
|||||
Description
ARL14EP Polyclonal specifically detects ARL14EP in Human samples. It is validated for Western Blot.Specifications
C11orf46 | |
Polyclonal | |
Rabbit | |
Q8N8R7 | |
120534 | |
Synthetic peptides corresponding to C11ORF46 The peptide sequence was selected from the N terminal of C11ORF46. Peptide sequence SSNDMLLLQLRTGMTLSGNNTICFHHVKIYIDRFEDLQKSCCDPFNIHKK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ADP-ribosylation factor-like 14 effector protein, ARF7EP, C11orf46, chromosome 11 open reading frame 46, dJ299F11.1, FLJ38968, hypothetical protein LOC120534 | |
ARL14EP | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title