Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001078 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001078 Supplier Novus Biologicals Supplier No. NB1001078
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARL2 Polyclonal specifically detects ARL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen ARL2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml, Flow Cytometry 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001658
Gene Alias ADP-ribosylation factor-like 2, ADP-ribosylation factor-like protein 2, ARFL2
Gene Symbols ARL2
Host Species Goat
Immunogen Peptide with sequence C-LDDISSRIFTAD corresponding to C-Terminus according to NP_001658.2.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 402
Test Specificity Human. Expected cross-reactivity based on sequence similarity: Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.