Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL2BP Antibody (OTI1A10), PerCP, Novus Biologicals™
SDP

Catalog No. NB271743PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271743PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271743PCP Supplier Novus Biologicals Supplier No. NBP271743PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARL2BP Monoclonal specifically detects ARL2BP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ARL2BP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1A10
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADP-ribosylation factor-like 2 binding protein, ADP-ribosylation factor-like protein 2-binding protein, ARF-like 2-binding protein, BART1binder of Arl Two, BARTArf-like 2 binding protein BART1, Binder of ARF2 protein 1, binder of Arl2
Gene Symbols ARL2BP
Host Species Mouse
Immunogen Full length recombinant protein of human ARL2BP (NP_036238) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23568.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.