Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL4 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001080 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001080 Supplier Novus Biologicals Supplier No. NB1001080
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARL4 Polyclonal specifically detects ARL4 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen ARL4
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P40617
Gene Alias ADP-ribosylation factor-like 4A, ADP-ribosylation factor-like protein 4A, ARL4ADP-ribosylation factor-like 4
Gene Symbols ARL4A
Host Species Goat
Immunogen Peptide with sequence GNGLSDQTSILSN-C corresponding to N-Terminus according to NP_001032241.1, NP_005729.1, NP_997625.1.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10124
Test Specificity This is also expected to react with a hypothetical human protein, XP_166703, and a hypothetical mouse protein, AAH03725 - both very similar to ARL4.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.