Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL5A Antibody, Novus Biologicals™
SDP

Catalog No. NBP15830420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15830420 20 μL
NBP158304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15830420 Supplier Novus Biologicals Supplier No. NBP15830420UL

Rabbit Polyclonal Antibody

ARL5A Polyclonal specifically detects ARL5A in Human samples. It is validated for Western Blot.

Specifications

Antigen ARL5A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9Y689
Gene Alias ADP-ribosylation factor-like 5, ADP-ribosylation factor-like 5A, ADP-ribosylation factor-like protein 5A, ARFLP5, ARL5ADP-ribosylation factor-like protein 5
Gene Symbols ARL5A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ARL5A(ADP-ribosylation factor-like 5A) The peptide sequence was selected from the middle region of ARL5A. Peptide sequence YKMLAHEDLRKAGLLIFANKQDVKECMTVAEISQFLKLTSIKDHQWHIQA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26225
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.