Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARL5B Antibody (OTI2D7), PerCP, Novus Biologicals™
SDP

Catalog No. NB272463PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272463PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272463PCP Supplier Novus Biologicals Supplier No. NBP272463PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARL5B Monoclonal specifically detects ARL5B in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ARL5B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2D7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADP-ribosylation factor-like 5B, ADP-ribosylation factor-like 8, ADP-ribosylation factor-like protein 5B, ADP-ribosylation factor-like protein 8, ADP-ribosylation-like factor 8, ARL8
Gene Symbols ARL5B
Host Species Mouse
Immunogen Full length recombinant protein of human ARL5B (NP_848930) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 221079.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.