Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Armenian Hamster anti-Integrin alpha 5/CD49e, Clone: HMa5-1, Novus Biologicals™
SDP

Catalog No. NBP1434015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1434015 0.5 mg
NBP1434015M 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1434015 Supplier Novus Biologicals Supplier No. NBP1434010.5MG

Armenian Hamster Monoclonal Antibody

Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Integrin alpha 5/CD49e
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone HMa5-1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
Gene Symbols ITGA5
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD49e.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.