Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARMER Antibody, Novus Biologicals™
SDP

Catalog No. NBP177198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177198 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177198 Supplier Novus Biologicals Supplier No. NBP177198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARMER Polyclonal specifically detects ARMER in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen ARMER
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 2 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q15041
Gene Alias ADP-ribosylation factor-like 6 interacting protein, ADP-ribosylation factor-like 6 interacting protein 1, Aip-1, AIP1, apoptotic regulator in the membrane of the endoplasmic reticulum, ARL-6-interacting protein 1, ARL6IPaip-1, ARMER, KIAA0069ADP-ribosylation factor-like protein 6-interacting protein 1
Gene Symbols ARL6IP1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the C-terminus of human ARMER. The immunogen is located within the last 50 amino acids of ARMER. Amino Acid Squence: KREINKLLKQKEKKNE
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 23204
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.