Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Novus Biologicals™ ARMER Recombinant Protein Antigen
 
    
    
    
        
            Shop All R&D Systems Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                0.1 mL
                            
                        
                    Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ARL6IP1. The ARMER Recombinant Protein Antigen is derived from E. coli. The ARMER Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
                        
                        
                        
                        
                        
                        
                        
                        
                            This is a blocking peptide for NBP1-91681. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifications
Specifications
| Gene ID (Entrez) | 23204 | 
| Species | Human | 
| Purification Method | Chromatography | 
| Purity | >80% | 
| Concentration | 0.5mg/mL | 
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. | 
| Formulation | PBS and 1M Urea, pH 7.4. | 
| For Use With (Application) | Blocking/Neutralizing, Control | 
| Gene Symbol | ARL6IP1 | 
| Label Type | Unlabeled | 
| Show More | 
For Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            