Learn More
PIP™ ArmorLite™ Full Nitrile Coated Cotton Gloves with Textured Finish
Shop All Protective Industrial Products Inc. ProductsDescription
- ActiFresh™ treated to reduce germs and odors
- Thinly coated with blue nitrile on cotton interlock liner
- Gloves are comfortable, flexible, and increase dexterity
- Textured coating is resistant to grease and oils for enhanced grip capabilities
- Effective, affordable protection from physical hazards such as cuts, punctures, snags and abrasion
- Knit Wrist helps prevent dirt and debris from entering the glove
- Made in ISO 9001:2000 / ISO 14001:2000 facilities
- Launderable
Specifications
Specifications
| Coated Area | Palm, Fingers |
| Coating Material | Nitrile |
| Color | Blue |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Product Type | Dip-coated Gloves |
| Size | Small |
| For Use With (Application) | General Handling, Assembly, Fabrication, Inspection, Maintenance, and Shipping/Receiving |
| Glove Cuff Color | Natural |
| Glove Cuff Style | Knit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.