Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ARN 14494

Click to view available options
Quantity:
10 mg
50 mg
Description
ARN 14494 is a potent serine palmitoyltransferase inhibitor (IC50 = 27.3 nM). ARN 14494 inhibits synthesis of long chain ceramides and dihydroceramides in an in vitro model of Alzheimer's diease. The compound prevents the synthesis of proinflammatory cytokines and the oxidative stress-related enzymes iNOS and COX-2 in mouse primary cortical astrocytes. ARN 14494 is neuroprotective against β-amyloid 1-42 induced neurotoxicity in primary cortical neurons co-cultured with astrocytes.
Specifications
Specifications
Chemical Name or Material | rel-1-[1-[2-[(3 aR,6 AS)-Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl]-2-oxoethyl]-4-piperidinyl]-2,3-dihydro-N-methyl-2-oxo-1 H-indole-5-carboxamide |
CAS | 1037834-43-7 |
Quantity | 10 mg |
Target | Other Transferase Inhibitors |
Molecular Formula | C24H32N4O3 |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction