Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNT/HIF-1 beta Antibody, Novus Biologicals™
SDP

Catalog No. NB100110SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100110SS 0.025 mL
NB100110 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100110SS Supplier Novus Biologicals Supplier No. NB100110SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 57 publications

ARNT/HIF-1 beta Polyclonal specifically detects ARNT/HIF-1 beta in Bovine, Canine, Ferret, Fish, Human, Mouse, Rat, Sheep samples. It is validated for Chemotaxis, ChIP Assay, EMSA, Gel Supershift, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen ARNT/HIF-1 beta
Applications ChIP Assay, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Simple Western 1:1000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:150, Immunoblotting, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. P27540
Gene Alias ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator
Gene Symbols ARNT
Host Species Rabbit
Immunogen Fusion protein to human HIF-1 beta containing amino acids 496-789. [UniProt# P27540]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 405
Test Specificity This is specific for HIF-1 beta/ARNT. It is not known if NB100-110 cross-reacts with ARNT2 which is related to HIF-1 beta/ARNT but is the product of a different gene.
Target Species Bovine, Mouse, Canine, Ferret, Fish, Fish, Human, Sheep, Mouse, Rat, Sheep
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.