Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARNT/HIF-1 beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 57 publications
$215.16 - $603.56
Specifications
| Antigen | ARNT/HIF-1 beta |
|---|---|
| Dilution | Western Blot 1:2000, Simple Western 1:1000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:150, Immunoblotting, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100110SS
![]() |
Novus Biologicals
NB100110SS |
0.025 mL |
Each for $215.16
|
|
|||||
NB100110
![]() |
Novus Biologicals
NB100110 |
0.1 mL |
Each for $603.56
|
|
|||||
Description
ARNT/HIF-1 beta Polyclonal specifically detects ARNT/HIF-1 beta in Human, Mouse, Rat, Bovine, Canine, Fish, Ferret, Sheep samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP).Specifications
| ARNT/HIF-1 beta | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators | |
| Whole antisera with 0.02% Sodium Azide | |
| ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator | |
| ARNT | |
| IgG | |
| Unpurified | |
| This is specific for HIF-1 beta/ARNT. It is not known if NB100-110 cross-reacts with ARNT2 which is related to HIF-1 beta/ARNT but is the product of a different gene. |
| Western Blot 1:2000, Simple Western 1:1000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:150, Immunoblotting, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Fish, Ferret, Sheep | |
| P27540 | |
| 405 | |
| Fusion protein to human HIF-1 beta containing amino acids 496-789. [UniProt# P27540] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title