Learn More
Description
Specifications
Specifications
| Antigen | ARNT/HIF-1 beta |
| Applications | Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), Gel Supershift, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | H1beta234 |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 25343232), Immunoprecipitation, Immunohistochemistry-Paraffin 1:100, Immunoblotting 1:100 - 1:2000, Gel Super Shift Assays reported in scientific literature (PMID 11325839), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing |
| Gene Accession No. | P27540 |
| Gene Alias | ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator |
| Gene Symbols | ARNT |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
