Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARNT/HIF-1 beta Antibody (H1beta234) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 96 publications
$243.50 - $560.50
Specifications
Antigen | ARNT/HIF-1 beta |
---|---|
Clone | H1beta234 |
Concentration | 1.4 mg/mL |
Dilution | Western Blot 1:500, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 25343232), Immunoprecipitation, Immunohistochemistry-Paraffin 1:100, Immunoblotting 1:100 - 1:2000, Gel Super Shift Assays reported in scientific literature (PMID 11325839), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100124SS
![]() |
Novus Biologicals
NB100124SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB100124
![]() |
Novus Biologicals
NB100124 |
0.1 mL |
Each for $560.50
|
|
|||||
Description
ARNT/HIF-1 beta Monoclonal specifically detects ARNT/HIF-1 beta in Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Gel Super Shift Assays, Chromatin Immunoprecipitation Sequencing.Specifications
ARNT/HIF-1 beta | |
1.4 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Angiogenesis, Autophagy, Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators | |
P27540 | |
405 | |
Fusion protein containing amino acids 496-789 of human HIF-1 beta. [Uniprot: P27540] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
H1beta234 | |
Western Blot 1:500, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 25343232), Immunoprecipitation, Immunohistochemistry-Paraffin 1:100, Immunoblotting 1:100 - 1:2000, Gel Super Shift Assays reported in scientific literature (PMID 11325839), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Chromatin Immunoprecipitation Sequencing | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep | |
ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator | |
ARNT | |
IgG1 κ | |
Protein G purified | |
92 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title