Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNT/HIF-1 beta Antibody (H1beta234), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB355151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355151 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355151 Supplier Novus Biologicals Supplier No. NB100124MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARNT/HIF-1 beta Monoclonal specifically detects ARNT/HIF-1 beta in Bovine, Ferret, Human, Mouse, Primate, Rat, Sheep samples. It is validated for ChIP Assay, ChIP sequencing (ChIP-seq), Gel Super Shift Assays, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen ARNT/HIF-1 beta
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone H1beta234
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Gel Super Shift Assays, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing
Formulation 50mM Sodium Borate
Gene Alias ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO
Host Species Mouse
Immunogen ARNT/HIF-1 beta Antibody (H1beta234) was developed against a fusion protein containing amino acids 496-789 of human HIF-1 beta. [Uniprot: P27540]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 405
Target Species Bovine, Ferret, Human, Mouse, Primate, Rat, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.