Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNTL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16900220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16900220 20 μL
NBP169002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16900220 Supplier Novus Biologicals Supplier No. NBP16900220UL

Rabbit Polyclonal Antibody

ARNTL2 Polyclonal specifically detects ARNTL2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ARNTL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q2VPD4
Gene Alias Aryl hydrocarbon receptor nuclear translocator-like protein 2, Basic-helix-loop-helix-PAS protein MOP9, bHLHe6, BMAL2, Brain and muscle ARNT-like 2, Class E basic helix-loop-helix protein 6, CLIF, CYCLE-like factor, Member of PAS protein 9, MOP9, PAS domain-containing protein 9, PASD9, Transcription Factor BMAL2
Gene Symbols ARNTL2
Host Species Rabbit
Immunogen The immunogen for anti-Arntl2 antibody: synthetic peptide corresponding to a region of Mouse ARNTL2 (NP_758513). Peptide sequence AILGYLPQELLGTSCYEYFHQDDHSSLTDKHKAVLQSKEKILTDSYKFRV.
Molecular Weight of Antigen 64 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56938
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.