Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNTL2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$501.50

Specifications

Antigen ARNTL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP169002
SDP
View Documents
Novus Biologicals
NBP169002
100 μL
Each for $501.50
Only null left
Add to Cart
 
NBP16900220
SDP
View Documents
Novus Biologicals
NBP16900220UL
20 μL N/A N/A N/A
Description

Description

ARNTL2 Polyclonal specifically detects ARNTL2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

ARNTL2
Polyclonal
Rabbit
Q2VPD4
56938
The immunogen for anti-Arntl2 antibody: synthetic peptide corresponding to a region of Mouse ARNTL2 (NP_758513). Peptide sequence AILGYLPQELLGTSCYEYFHQDDHSSLTDKHKAVLQSKEKILTDSYKFRV.
Primary
Western Blot
Unconjugated
RUO
Aryl hydrocarbon receptor nuclear translocator-like protein 2, Basic-helix-loop-helix-PAS protein MOP9, bHLHe6, BMAL2, Brain and muscle ARNT-like 2, Class E basic helix-loop-helix protein 6, CLIF, CYCLE-like factor, Member of PAS protein 9, MOP9, PAS domain-containing protein 9, PASD9, Transcription Factor BMAL2
ARNTL2
IgG
64 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.