Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNTL2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10075238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075238 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075238 Supplier Novus Biologicals Supplier No. NB10075238
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ARNTL2 Polyclonal specifically detects ARNTL2 in Human samples. It is validated for Western Blot.

Specifications

Antigen ARNTL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with No Preservative
Gene Accession No. Q9H2M4
Gene Alias Aryl hydrocarbon receptor nuclear translocator-like protein 2, Basic-helix-loop-helix-PAS protein MOP9, bHLHe6, BMAL2, Brain and muscle ARNT-like 2, Class E basic helix-loop-helix protein 6, CLIF, CYCLE-like factor, Member of PAS protein 9, MOP9, PAS domain-containing protein 9, PASD9, Transcription Factor BMAL2
Gene Symbols ARNTL2
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 8a.a. to 100a.a. from ARNTL2(a 551a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56938
Test Specificity ARNTL2
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.