Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aromatase Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB060174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060174 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060174 Supplier Novus Biologicals Supplier No. NBP299350DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aromatase Polyclonal specifically detects Aromatase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Aromatase
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Aromatase.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1588
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.