Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARPC3 Antibody, Novus Biologicals™
SDP

Catalog No. NB300895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300895 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300895 Supplier Novus Biologicals Supplier No. NB300895
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARPC3 Polyclonal specifically detects ARPC3 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ARPC3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. O15145
Gene Alias actin related protein 2/3 complex, subunit 3, 21kDa, subunit 3 (21 kD)
Gene Symbols ARPC3
Host Species Goat
Immunogen Peptide with sequence C-RQFMNKSLSGPGQ corresponding to C-Terminus according to NP_005710.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10094
Test Specificity Please note there are two hypothetical proteins called “similar to the Arp2/3 protein complex subunit p21-Arc” that are virtually identical (CAC14083, XP_208062).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.