Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Arsenal™ 5020 Standard Polyester Gear Duffel Bag
Arsenal™ 5020 Standard Gear Duffel Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.
$44.13 - $89.66
Specifications
| Material | 600D Polyester |
|---|---|
| For Use With (Application) | Fire and Rescue, EMTs, Construction, General Purpose |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Color | Volume (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Volume (Metric) | Price | Quantity & Availability | ||||
|
19-819-806
|
Ergodyne™
13319 |
Black | 24 L |
Each for $44.13
|
|
||||
|
19-819-807
|
Ergodyne™
13320 |
Black | 54 L |
Each for $51.89
|
|
||||
|
19-819-808
|
Ergodyne™
13321 |
Black | 79 L |
Each for $89.66
|
|
||||
|
19-819-812
|
Ergodyne™
13322 |
Black | 112 L |
Each for $64.34
|
|
||||
|
19-819-826
|
Ergodyne™
13419 |
Realtree Xtra™ | 24 L | N/A | N/A | N/A | |||
|
19-819-827
|
Ergodyne™
13420 |
Realtree Xtra™ | 54 L | N/A | N/A | N/A | |||
|
19-819-828
|
Ergodyne™
13421 |
Realtree Xtra™ | 79 L | N/A | N/A | N/A | |||
|
19-819-829
|
Ergodyne™
13422 |
Realtree Xtra™ | 112 L | N/A | N/A | N/A | |||
Description
- Durable material withstands wear and tear
- Water-resistant backing helps to repel water and stains
- Heavy-duty webbing straps on top and side of duffel for extra reinforcement
- Removable and adjustable padded shoulder strap for easy transportation
- Heavy-duty zippers to secure gear inside the duffel
- Two end accessory pockets for cell phones, headphones, pens, keys, and other small parts
Specifications
| 600D Polyester | |
| Fire and Rescue, EMTs, Construction, General Purpose | |
| 1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title