Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5030 Water Resistant Duffel Bag

Weather resistant duffel bag with maximum storage capacity is constructed of tarpaulin fabric to withstand hard weather conditions.

$192.95 - $242.69

Specifications

Color Black
Material 1000D Poly Mesh Tarpaulin
For Use With (Application) General Purpose
Quantity 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
19-819-739
View Documents
Ergodyne™
13030
51 L
Each for $192.95
Only null left
Add to Cart
 
19-819-740
View Documents
Ergodyne™
13032
72 L
Each for $201.41
Only null left
Add to Cart
 
19-819-745
View Documents
Ergodyne™
13034
112 L
Each for $242.69
Only null left
Add to Cart
 
Description

Description

  • Water-resistant tarpaulin with 1000D poly mesh
  • Shoulder straps carry as duffel or backpack
  • Four compression straps and two haul handles
  • Open design for maximum storage capacity
  • ID and document pockets on top of bag
Specifications

Specifications

Black
1000D Poly Mesh Tarpaulin
General Purpose
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.