Learn More
Arsenic(III) oxide, Puratronic™, 99.996% (metals basis excluding Sb), Sb content reported
Description
Arsenic trioxide is used as a precursor to forestry products, in colorless glass production, and in electronics. And it is also used as pigments, and preservatives for hides and wood, in the semiconductor industry to create light emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Boiling Point | 465°C (sublimation) | 
| Melting Point | 312.3°C | 
| Physical Form | Powder | 
| Quantity | 2 g | 
| UN Number | UN1561 | 
| Merck Index | 14,804 | 
| Solubility Information | Soluble in hot water,dilute acids and alkalies. Insoluble in alcohol,chloroform,and ether,. | 
| Formula Weight | 197.84 | 
| Percent Purity | 99.996% | 
| Grade | Puratronic™ | 
| Show More | 
Safety and Handling
| Hazard Category | H300-H314-H335-H350-H371 | 
| Hazard Statement | GHS H Statement H300-H350-H314-H318 Fatal if swallowed. May cause cancer. Causes severe skin burns and eye damage. Causes serious eye damage. | 
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c | 
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC TRIOXIDE | 
| EINECSNumber | 215-481-4 | 
| RTECSNumber | CG3325000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                