Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) sulfide, 99.999% (metals basis)

Catalog No. p-7051787
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4198006 5 g
AA4198014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4198006 Supplier Thermo Scientific Chemicals Supplier No. 04198006
Only null left
Add to Cart

For vacuum deposition | CAS: 1303-33-9 | As2S3 | 246.02 g/mol

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]

Specifications

Boiling Point 707°C
Melting Point 300°C
Physical Form Fused Lump
Quantity 5 g
UN Number UN1557
Merck Index 14,806
Formula Weight 246.03
Percent Purity 99.999%
Odor Like rotten eggs
Density 3.43 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic(III) sulfide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 215-117-4
RTECSNumber CG2638000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.