Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Trioxide, ACS Reagent (Arsenic (III) Oxide, Reductometric Standard), Reagents

Catalog No. 01338074
Change view
Click to view available options
:
Quantity: 125 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-074 Quantity: 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-074 Supplier Reagents Holdings Llc Supplier No. C1143500125D2
Only null left
Add to Cart
Add to Cart

Arsenic Trioxide, ACS Reagent (Arsenic (III) Oxide, Reductometric Standard), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1327-53-3
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
Synonym Arsenic(III) Oxide
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Boiling Point 465°C
Melting Point 193°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.738
Chemical Name or Material Arsenic Trioxide, ACS Reagent (Arsenic (III) Oxide, Reductometric Standard)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.