Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ART1 Antibody (OTI6E2), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N270211A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270211A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270211A647 Supplier Novus Biologicals Supplier No. NBP270211AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ART1 Monoclonal specifically detects ART1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ART1
Applications Western Blot
Classification Monoclonal
Clone OTI6E2
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6
Gene Symbols ART1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 83-295 of human ART1 (NP_004305) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 417.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.