Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ART2.2/Art2b Antibody (Nika102), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10065853V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065853V 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065853V Supplier Novus Biologicals Supplier No. NB10065853V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

ART2.2/Art2b Monoclonal specifically detects ART2.2/Art2b in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen ART2.2/Art2b
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Nika102
Conjugate DyLight 405
Dilution Flow Cytometry
Gene Alias ADP-ribosyltransferase 2b, Rt6, Rt-6, Rt6.2, Rt6-2
Gene Symbols ART2B
Host Species Rat
Immunogen pME.CD8LF-ART2.2
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11872
Test Specificity NB100-65853 recognizes the gene product of the mouse ADP-ribosyltransferase (ART) 2.2 gene, an ortholog of rat RT6. ART2.2 expressed exclusively by mature T-cells. Levels of ART2.2 expression vary between inbred strains of mice and is absent in NZW mice, in which the ART2.2 gene is deleted (PMID: 10570289, Koch-Nolte et al. 1999).
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.