Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ART3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB064606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064606 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064606 Supplier Novus Biologicals Supplier No. NBP300144F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ART3 Polyclonal specifically detects ART3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ART3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ADP-ribosyltransferase 3, EC 2.4.2.31, ecto-ADP-ribosyltransferase 3, FLJ26404, Mono(ADP-ribosyl)transferase 3, mono-ADP-ribosyltransferase, NAD(P)(+)--arginine ADP-ribosyltransferase 3, TMART
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ART3 (AAH08397.1; Met1-Ser362).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 419
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.