Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemin Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB360790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360790 Supplier Novus Biologicals Supplier No. NBP223618F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Artemin Polyclonal antibody specifically detects Artemin in Human samples. It is validated for Western Blot

Specifications

Antigen Artemin
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin
Host Species Rabbit
Immunogen Reacts with the N terminus of Artemin.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9048
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.