Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemin Antibody (RM0074-6G32), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB362046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362046 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362046 Supplier Novus Biologicals Supplier No. NBP121701MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Artemin Monoclonal antibody specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen Artemin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0074-6G32
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin
Host Species Rat
Immunogen Mouse recombinant Artemin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9048
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.