Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemis Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000931 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000931 Supplier Novus Biologicals Supplier No. NB100183SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Artemis Polyclonal specifically detects Artemis in Human samples. It is validated for Western Blot.

Specifications

Antigen Artemis
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias ASCID, DCLREC1C, DNA cross-link repair 1C, DNA cross-link repair 1C protein, EC 3.1, FLJ36438, hSNM1C, protein artemis, Protein A-SCID, PSO2 homolog, RS-SCID, severe combined immunodeficiency, type a (Athabascan), SNM1 homolog C, SNM1CS. cerevisiae), SNM1-like protein
Gene Symbols DCLRE1C
Host Species Rabbit
Immunogen A synthetic peptide made to residues 677-692 [GESIAVKKRKCSLLDT] of the human Artemis protein.
Molecular Weight of Antigen 66 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Cancer, DNA Repair, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 64421
Test Specificity Artemis
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.