Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arylsulfatase A/ARSA Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB326137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326137 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326137 Supplier Novus Biologicals Supplier No. NBP299307MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Arylsulfatase A/ARSA Polyclonal antibody specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Arylsulfatase A/ARSA
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Arylsulfatase A/ARSA (Uniprot#: P15289-1; Met 1-Ala 507).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 410
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.