Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arylsulfatase A/ARSA Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
SDP

Catalog No. NB022161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022161 100 μg
NB022162 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB022161 Supplier Novus Biologicals Supplier No. NBP290354100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Arylsulfatase A/ARSA Monoclonal specifically detects Arylsulfatase A/ARSA in Mouse samples. It is validated for ELISA.

Specifications

Antigen Arylsulfatase A/ARSA
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD
Host Species Rabbit
Immunogen Recombinant Mouse Arylsulfatase A/ARSA protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 410
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.