Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arylsulfatase D Antibody, Novus Biologicals™
SDP

Catalog No. NBP120886 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120886 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120886 Supplier Novus Biologicals Supplier No. NBP120886
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Arylsulfatase D Polyclonal specifically detects Arylsulfatase D in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Arylsulfatase D
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001660.2
Gene Alias arylsulfatase D, ASD, EC 3.1.6.-, EC 3.1.6.2
Gene Symbols ARSD
Host Species Goat
Immunogen Peptide with sequence C-DASNGYRALQWNA corresponding to internal region according to NP_001660.2, NP_033667.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 414
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001660.2; NP_033667.2)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.