Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Tocris Bioscience™ AS 1842856
    
    
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                10 mg
                            
                        
                            
                                50 mg
                            
                        
                    Description
AS 1842856 is a potent and selective Foxo1 inhibitor (IC50 values are 33 nM at Foxo1 and >1 μM at Foxo3a and Foxo4); decreases Foxo-mediated promoter activity by 70%, 20% and 3% at Foxo1, Foxo4 and Foxo3, respectively. Regulates gluconeogenesis both in vitro and in vivo. Attenuates fasting plasma glucose level in diabetic db/db mice. Also suppresses autophagy, adipocyte differentiation and soluble guanylyl cyclase expression. Also inhibits STAT3 signaling in NSCLC. Orally bioavailable.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| Chemical Name or Material | 5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid | 
| CAS | 836620-48-5 | 
| Quantity | 10 mg | 
| Molecular Formula | C18H22FN3O3 | 
| Purity | 98% | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction