Learn More
ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, Biotin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | ASAH2/N-acylsphingosine Amidohydrolase-2 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, NCDase, N-CDase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant ASAH2/N-acylsphingosine Amidohydrolase-2 (NP_061300.1). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.