Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358352 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358352 Supplier Novus Biologicals Supplier No. NBP176360MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal antibody specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen ASAH2/N-acylsphingosine Amidohydrolase-2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, NCDase, N-CDase
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human ASAH2. The immunogen is located within the last 50 amino acids of ASAH2. Amino Acid Squence: YRIRYFGHNRKQDILK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56624
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.