Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAH3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1742520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1742520 20 μL
NBP174125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1742520 Supplier Novus Biologicals Supplier No. NBP17412520UL

Rabbit Polyclonal Antibody

ASAH3 Polyclonal specifically detects ASAH3 in Human samples. It is validated for Western Blot.

Specifications

Antigen ASAH3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8TDN7
Gene Alias Acylsphingosine deacylase 3, Alkaline CDase 1, alkaline ceramidase 1, AlkCDase 1, ALKCDase1, ASAH3, EC 3.5.1.23, MGC138327, MGC138329, N-acylsphingosine amidohydrolase (alkaline ceramidase) 3, N-acylsphingosine amidohydrolase 3
Gene Symbols ACER1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the C terminal of ACER1. Immunizing peptide sequence ITFPYGMVTMALVDANYEMPGETLKVRYWPRDSWPVGLPYVEIRGDDKDC.
Molecular Weight of Antigen 31 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125981
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.