Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAM Antibody (001), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB075876 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075876 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075876 Supplier Novus Biologicals Supplier No. NBP290634IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ASAM Monoclonal specifically detects ASAM in Mouse samples. It is validated for ELISA.

Specifications

Antigen ASAM
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias ACAMCoxsackie- and adenovirus receptor-like membrane protein, Adipocyte adhesion molecule, adipocyte-specific adhesion molecule, ASAMCAR-like membrane protein, CXADR-like membrane protein, FLJ22415
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASAM (Q8R373-1; Met 1-Met 232).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79827
Test Specificity No cross-reactivity in ELISA with: Human ASAM
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.