Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAP1, p Tyr782 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177933 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177933 Supplier Novus Biologicals Supplier No. NBP177933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASAP1 Polyclonal specifically detects ASAP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ASAP1 (p Tyr782)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1.09 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:4000-1:16000, Immunohistochemistry 20-40 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 20 ug/ml, Immunohistochemistry-Frozen
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. NP_034156, Q9QWY8
Gene Alias 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activatingprotein, AMAP1, ARF GTPase-activating protein 1, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1, ArfGAP with SH3 domain, ankyrin repeat and PH domain 1, centaurin, beta 4, CENTB4, DDEF1ADP-ribosylation factor-directed GTPase-activating protein 1, development and differentiation enhancing factor 1, Development and differentiation-enhancing factor 1, Differentiation-enhancing factor 1, KIAA1249DEF-1, PAG2, PAP, PIP2-dependent ARF1 GAP, ZG14P
Gene Symbols ASAP1
Host Species Rabbit
Immunogen This affinity purified ASAP1 [p Tyr782] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 775-800 of mouse ASAP1 protein. (Uniprot: Q9QWY8)
Molecular Weight of Antigen 127.42 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 50807
Test Specificity This affinity-purified antibody is directed against the phosphorylated form of mouse ASAP1 protein at the pY782 residue. The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against mouse ASAP1 pY782 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated mouse ASAP1 is minimal by ELISA. A BLAST analysis was used to suggest cross reactivity with ASAP1 proteins from human, chicken, bovine, dog, rat and chimpanzee based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Chimpanzee
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.